3-Chloro-6-methoxyisoquinoline 97%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-6-methoxy-2-azanaphthalene

A411298-100MG A411298-250MG
77278-882EA 47 USD
77278-882 77278-884
3-Chloro-6-methoxyisoquinoline 97%
3-Chloro-6-methoxyisoquinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClNO
MW: 193.63 g/mol
MDL Number: MFCD11044725
CAS Number: 82117-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR