4,4'-Methylenebis(2-ethylaniline) 98%

Supplier: Ambeed
Warning

A410855-100G A410855-25G A410855-500G
77291-294EA 18.54 USD
77291-294 77291-296 77291-298
4,4'-Methylenebis(2-ethylaniline) 98%
4,4'-Methylenebis(2-ethylaniline)

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂
MW: 254.37 g/mol
MDL Number: MFCD00128916
CAS Number: 19900-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR