3,5-Dimethylbenzyl alcohol 98%

Supplier: Ambeed
Warning

A410450-1G A410450-25G A410450-5G
77278-860EA 9 USD
77278-860 77278-862 77278-864
3,5-Dimethylbenzyl alcohol 98%
3,5-Dimethylbenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00004648
CAS Number: 27129-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR