2,5-Diaminohydroquinone dihydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 2,5-Diaminobenzene-1,4-diol dihydrochloride

A410260-100MG A410260-1G A410260-250MG A410260-5G
77252-206EA 18.54 USD
77252-206 77252-208 77252-210 77252-212
2,5-Diaminohydroquinone dihydrochloride 95%
2,5-Diaminohydroquinone dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₀Cl₂N₂O₂
MW: 213.06 g/mol
MDL Number: MFCD00239416
CAS Number: 24171-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR