4-Ethylbenzaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: p-Ethylbenzaldehyde

A410136-25G A410136-5G A410136-100G
77291-276EA 25.75 USD
77291-276 77291-278 77616-782
4-Ethylbenzaldehyde 98%
4-Ethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O
MW: 134.18 g/mol
MDL Number: MFCD00006956
CAS Number: 4748-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR