4-(Difluoromethoxy)phenylacetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(Difluoromethoxy)phenyl)acetonitrile

A409964-1G A409964-250MG A409964-25G A409964-5G
77264-574EA 21.63 USD
77264-574 77264-576 77264-578 77264-580
4-(Difluoromethoxy)phenylacetonitrile 98%
4-(Difluoromethoxy)phenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₇F₂NO
MW: 183.1600 g/mol
MDL Number: MFCD00236242
CAS Number: 41429-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR