N-Cyclohexyl-2-benzothiazolylsulfenamide 95%

Supplier: Ambeed
Danger

A409501-25G A409501-100G A409501-500G A409501-1000G
77240-732EA 5 USD
77240-732 77616-776 77616-778 77746-827
N-Cyclohexyl-2-benzothiazolylsulfenamide 95%
N-Cyclohexyl-2-benzothiazolylsulfenamide

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂S₂
MDL Number: MFCD00022872
CAS Number: 95-33-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR