3-Methoxy-2-nitrobenzonitrile 95%

Supplier: Ambeed
Warning

A409408-1G A409408-250MG A409408-5G
77278-854EA 49.44 USD
77278-854 77278-856 77278-858
3-Methoxy-2-nitrobenzonitrile 95%
3-Methoxy-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₈H₆N₂O₃
MW: 178.14 g/mol
MDL Number: MFCD18382535
CAS Number: 142596-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR