Dibenzo[b,d]thiophen-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Aminodibenzothiophene

A409275-100MG A409275-1G A409275-250MG A409275-5G
77240-724EA 6.18 USD
77240-724 77240-726 77240-728 77240-730
Dibenzo[b,d]thiophen-2-amine 97%
Dibenzo[b,d]thiophen-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₉NS
MW: 199.27 g/mol
MDL Number: MFCD00995227
CAS Number: 7428-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR