3'-Brom-4'-methyloacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 3'-Bromo-4'-methylacetophenone, 1-(3-Bromo-4-methylphenyl)ethanone

A408626-100G A408626-10G A408626-1G A408626-25G A408626-5G
77264-560EA 255.44 USD
77264-560 77264-562 77264-564 77264-566 77264-568
3'-Brom-4'-methyloacetophenone 98%
3'-Brom-4'-methyloacetophenone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MW: 213.0700 g/mol
MDL Number: MFCD02683854
CAS Number: 40180-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR