3,5-Dichloropyridine-2-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine

A408326-100G A408326-10G A408326-1G A408326-25G A408326-500G
77278-840EA 127 USD
77278-840 77278-842 77278-844 77278-846 77278-848
3,5-Dichloropyridine-2-carbonitrile 97%
3,5-Dichloropyridine-2-carbonitrile

Precaución: for Research Use Only

Formula: C₆H₂Cl₂N₂
MDL Number: MFCD03788758
CAS Number: 85331-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR