1,3,5,7-Tetrabromoadamantane 97%

Supplier: Ambeed
Warning

A408286-1G A408286-250MG A408286-10G
77240-714EA 34 USD
77240-714 77240-716 77746-824
1,3,5,7-Tetrabromoadamantane 97%
1,3,5,7-Tetrabromoadamantane

Precaución: for Research Use Only

Formula: C₁₀H₁₂Br₄
MDL Number: MFCD00193968
CAS Number: 7314-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR