4-Bromo-3,5-dimethoxybenzoic acid 97%

Supplier: Ambeed
Warning

A408274-10G A408274-1G A408274-25G A408274-5G
77278-836EA 19.57 USD
77278-836 77278-838 77748-825 77748-826
4-Bromo-3,5-dimethoxybenzoic acid 97%
4-Bromo-3,5-dimethoxybenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₄
MW: 261.07 g/mol
MDL Number: MFCD01632140
CAS Number: 56518-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR