4-(Isobutylcarbamoyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (4-(Isobutylcarbamoyl)phenyl)boronic acid, 4-(Isobutylaminocarbonyl)phenylboronic acid, 4-(Isobutylcarbamoyl)benzeneboronic acid

A407682-1G A407682-5G
77291-260EA 70.04 USD
77291-260 77291-262
4-(Isobutylcarbamoyl)phenylboronic acid 98%
4-(Isobutylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MW: 221.06 g/mol
MDL Number: MFCD03411955
CAS Number: 850568-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR