tert-Butyl 3-(piperazin-1-yl)azetidine-1-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl 3-piperazin-1-ylazetidine-1-carboxylate

A407555-1G A407555-250MG A407555-100MG A407555-5G
77240-704EA 291.49 USD
77240-704 77240-706 77616-772 77746-821
tert-Butyl 3-(piperazin-1-yl)azetidine-1-carboxylate 97%
tert-Butyl 3-(piperazin-1-yl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₃N₃O₂
MW: 241.33 g/mol
MDL Number: MFCD09264407
CAS Number: 178311-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR