4,6-Dimethyl-2,3-dihydro-1H-inden-1-one 97%

Supplier: Ambeed
Warning

A407481-250MG A407481-1G A407481-5G
77252-186EA 25 USD
77252-186 77746-818 77746-819
4,6-Dimethyl-2,3-dihydro-1H-inden-1-one 97%
4,6-Dimethyl-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₁H₁₂O
MDL Number: MFCD09908137
CAS Number: 1685-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR