5''-(4'-Amino-[1,1'-biphenyl]-4-yl)-[1,1':4',1'':3'',1''':4''',1''''-quinquephenyl]-4,4''''-diamine 98%

Supplier: Ambeed
Warning

A407404-250MG A407404-5G
77264-552EA 42.23 USD
77264-552 77264-554
5''-(4'-Amino-[1,1'-biphenyl]-4-yl)-[1,1':4',1'':3'',1''':4''',1''''-quinquephenyl]-4,4''''-diamine 98%
5''-(4'-Amino-[1,1'-biphenyl]-4-yl)-[1,1':4',1'':3'',1''':4''',1''''-quinquephenyl]-4,4''''-diamine

Precaución: for Research Use Only

Formula: C₄₂H₃₃N₃
MW: 579.73 g/mol
MDL Number: MFCD32667108
CAS Number: 1400987-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR