GNE-493 98+%

Supplier: Ambeed
Warning

Synonyms: 2-(2-(2-Aminopyrimidin-5-yl)-4-morpholinothieno[3,2-d]pyrimidin-6-yl)propan-2-ol

A407156-10MG A407156-1MG A407156-5MG
77240-692EA 101.97 USD
77240-692 77240-694 77240-696
GNE-493 98+%
GNE-493

Precaución: for Research Use Only

Formula: C₁₇H₂₀N₆O₂S
MW: 372.4 g/mol
MDL Number: MFCD18251526
CAS Number: 1033735-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR