4-Fluoro-ɑ-methylstyrene 98% stabilized

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-4-(1-methylethenyl)benzene, 2-(4-Fluorophenyl)-1-propene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A406760-10G A406760-1G A406760-25G A406760-5G
77291-238EA 42.23 USD
77291-238 77291-240 77291-242 77291-244
4-Fluoro-ɑ-methylstyrene 98% stabilized
4-Fluoro-alpha-methylstyrene

Precaución: for Research Use Only

Formula: C₉H₉F
MW: 136.17 g/mol
MDL Number: MFCD00042297
CAS Number: 350-40-3
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR