Triethoxy(pentafluorophenyl)silane 98%

Supplier: Ambeed
Warning

Synonyms: (Triethoxysilyl)pentafluorobenzene, Triethoxy(perfluorophenyl)silane

A406635-100MG A406635-1G A406635-250MG A406635-5G
77252-170EA 13.39 USD
77252-170 77252-172 77252-174 77252-176
Triethoxy(pentafluorophenyl)silane 98%
Triethoxy(pentafluorophenyl)silane

Precaución: for Research Use Only

Formula: C₁₂H₁₅F₅O₃Si
MW: 330.33 g/mol
MDL Number: MFCD03411257
CAS Number: 20083-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR