3-(3-Nitrophenyl)isoxazol-5-amine 95%

Supplier: Ambeed
Warning

A406391-10G A406391-1G A406391-5G
77278-830EA 365.65 USD
77278-830 77278-832 77278-834
3-(3-Nitrophenyl)isoxazol-5-amine 95%
3-(3-Nitrophenyl)isoxazol-5-amine

Precaución: for Research Use Only

Formula: C₉H₇N₃O₃
MW: 205.17 g/mol
MDL Number: MFCD07383079
CAS Number: 887591-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR