7-Acetoxy-4-methylcoumarin 98%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-2-oxo-2H-chromen-7-yl acetate, 4-Methylumbelliferyl acetate

A406230-1G A406230-5G
77291-234EA 9.27 USD
77291-234 77291-236
7-Acetoxy-4-methylcoumarin 98%
7-Acetoxy-4-methylcoumarin

Precaución: for Research Use Only

Formula: C₁₂H₁₀O₄
MW: 218.21 g/mol
MDL Number: MFCD00006865
CAS Number: 2747-05-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR