Bromo-PEG6-alcohol 97%

Supplier: Ambeed
Warning

Synonyms: 17-Bromo-3,6,9,12,15-pentaoxaheptadecan-1-ol

A406203-100MG A406203-1G A406203-250MG A406203-5G
77264-550EA 17 USD
77264-550 77616-766 77616-768 77616-770
Bromo-PEG6-alcohol 97%
Bromo-PEG6-alcohol

Precaución: for Research Use Only

Formula: C₁₂H₂₅BrO₆
MDL Number: MFCD20926394
CAS Number: 136399-05-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR