3-Amino-2-bromobenzoic acid 97%

Supplier: Ambeed
Danger

A406178-1G A406178-5G A406178-25G
77278-828EA 21 USD
77278-828 77748-823 77748-822
3-Amino-2-bromobenzoic acid 97%
3-Amino-2-bromobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MDL Number: MFCD07786957
CAS Number: 168899-61-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR