3-bromo-4-methoxybenzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-4-methoxybenzene-1-sulphonyl chloride

A405327-100MG A405327-1G A405327-250MG A405327-5G
77278-806EA 6 USD
77278-806 77278-808 77278-810 77278-812 77278-814
3-bromo-4-methoxybenzenesulfonyl chloride 98%
3-bromo-4-methoxybenzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₆BrClO₃S
MDL Number: MFCD03989643
CAS Number: 23094-96-4
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR