(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine 98%

Supplier: Ambeed
Warning

A405325-100MG A405325-1G A405325-250MG A405325-50MG
77252-162EA 20 USD
77252-162 77252-164 77252-166 77252-168
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine 98%
(R)-(-)-1-[(S)-2-(Dicyclohexylphosphino)ferrocenyl]ethyldiphenylphosphine

Precaución: for Research Use Only

Formula: C₃₆H₄₄FeP₂
MDL Number: MFCD00800280
CAS Number: 158923-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR