1-(2-Chloro-4-morpholinothieno[3,2-d]pyrimidin-6-yl)-N-methylmethanamine 97%

Supplier: Ambeed
Warning

A404852-100MG A404852-1G
77240-682EA 70.04 USD
77240-682 77240-684
1-(2-Chloro-4-morpholinothieno[3,2-d]pyrimidin-6-yl)-N-methylmethanamine 97%
1-(2-Chloro-4-morpholinothieno[3,2-d]pyrimidin-6-yl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₅ClN₄OS
MW: 298.79 g/mol
MDL Number: MFCD28966047
CAS Number: 955979-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR