(S)-4-Isopropyloxazolidine-2-thione 95+%

Supplier: Ambeed
Warning

A404500-1G A404500-250MG
77291-214EA 98.88 USD
77291-214 77291-216
(S)-4-Isopropyloxazolidine-2-thione 95+%
(S)-4-Isopropyloxazolidine-2-thione

Precaución: for Research Use Only

Formula: C₆H₁₁NOS
MW: 145.22 g/mol
MDL Number: MFCD00051949
CAS Number: 104499-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR