1H-Pyrrolo[3,2-b]pyridine-6-carbonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 6-Cyano-4-azaindole

A404156-100MG A404156-1G A404156-250MG A404156-25G A404156-5G
77240-666EA 31.93 USD
77240-666 77240-668 77240-670 77240-672 77240-674
1H-Pyrrolo[3,2-b]pyridine-6-carbonitrile 95%
1H-Pyrrolo[3,2-b]pyridine-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₅N₃
MW: 143.15 g/mol
MDL Number: MFCD09763664
CAS Number: 944937-79-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR