2-Acetoxybenzonitrile 97%

Supplier: Ambeed
Warning

A404111-1G A404111-25G A404111-5G
77252-144EA 20 USD
77252-144 77252-146 77252-148
2-Acetoxybenzonitrile 97%
2-Acetoxybenzonitrile

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD00017360
CAS Number: 5715-02-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR