4,4'-Ethylenedianiline 98%

Supplier: Ambeed
Warning

Synonyms: 4,4'-Diamino-1,2-diphenylethane, 4,4'-Diaminobibenzyl, alpha,alpha'-Bi-p-toluidine, 4,4'-(Ethane-1,2-diyl)dianiline

A404004-100G A404004-1G A404004-5G
77291-194EA 916.7 USD
77291-194 77291-196 77291-198
4,4'-Ethylenedianiline 98%
4,4'-Ethylenedianiline

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂
MW: 212.30 g/mol
MDL Number: MFCD00007923
CAS Number: 621-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR