[RuCl(p-cymene)((S)-xylbinap)]Cl 98%

Supplier: Ambeed
Warning

Synonyms: Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}(p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-xylbinap)]Cl, Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}(p-cymene)ruthenium(II) chloride, Chloro[(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride

A403945-100MG A403945-1G A403945-250MG
77252-138EA 52 USD
77252-138 77252-140 77252-142
[RuCl(p-cymene)((S)-xylbinap)]Cl 98%
[RuCl(p-cymene)((S)-xylbinap)]Cl

Precaución: for Research Use Only

Formula: C₆₂H₆₂Cl₂P₂Ru
MW: 1041.10 g/mol
MDL Number: MFCD09753015
CAS Number: 944451-25-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR