Fmoc-Tic-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-L-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, (S)-2-(((9H-Fluoren-9-yl)methoxy)carbonyl)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid

A403656-10G A403656-1G A403656-25G A403656-5G
77291-190EA 16 USD
77291-190 77291-192 77572-252 77594-224
Fmoc-Tic-OH 98%
Fmoc-Tic-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₁NO₄
MW: 399.446 g/mol
MDL Number: MFCD00144368
CAS Number: 136030-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR