1H-Pyrrole-2-carboxamide 95%

Supplier: Ambeed
Warning

Synonyms: 2-Carbamoyl-1H-pyrrole, 2-(Aminocarbonyl)-1H-pyrrole

A403611-1G A403611-250MG
77240-660EA 120.51 USD
77240-660 77240-662
1H-Pyrrole-2-carboxamide 95%
1H-Pyrrole-2-carboxamide

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.114 g/mol
MDL Number: MFCD00273121
CAS Number: 4551-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR