1,2-Dimethyl-2,3-dihydro-1H-indole-5-carbaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dimethylindoline-5-carbaldehyde

A403485-100MG A403485-1G A403485-250MG
77240-654EA 8 USD
77240-654 77240-656 77240-658
1,2-Dimethyl-2,3-dihydro-1H-indole-5-carbaldehyde 97%
1,2-Dimethyl-2,3-dihydro-1H-indole-5-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD07790518
CAS Number: 887690-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR