3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-tert-Butyl-1-methylpyrazole-5-carboxylic Acid

A403367-10G A403367-250MG A403367-5G A403367-1G
77278-774EA 165.83 USD
77278-774 77278-776 77278-778 77748-811
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid 97%
3-(tert-Butyl)-1-methyl-1H-pyrazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O₂
MW: 182.22 g/mol
MDL Number: MFCD00173812
CAS Number: 175277-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR