ω-Bromo-2-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: α-Bromo-2-nitroacetophenone, 2-Bromo-2'-nitroacetophenone, Bromomethyl o-nitrophenyl ketone, 2-Nitrophenacyl bromide

A402937-100G A402937-10G A402937-1G A402937-25G A402937-5G
77240-644EA 195 USD
77240-644 77240-646 77240-648 77240-650 77240-652
ω-Bromo-2-nitroacetophenone 98%
ω-Bromo-2-nitroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆BrNO₃
MDL Number: MFCD00010294
CAS Number: 6851-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR