2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-(Cyclopropylmethoxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 6-(Cyclopropylmethoxy)pyridine-3-boronic acid pinacol ester

A402438-1G A402438-250MG A402438-5G
77240-630EA 124.63 USD
77240-630 77240-632 77240-634
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester 97%
2-(Cyclopropylmethoxy)-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₂BNO₃
MW: 275.16 g/mol
MDL Number: MFCD07781184
CAS Number: 947191-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR