2-Phenyl-1H-imidazole-4-carbaldehyde 95%
Supplier: Ambeed
|
Warning
|
A402242-1G
A402242-250MG
A402242-5G
77278-746EA
89.61
USD
77278-746
77278-748
77278-750
2-Phenyl-1H-imidazole-4-carbaldehyde 95%
2-Phenyl-1H-imidazole-4-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₈N₂O MW: 172.19 g/mol |
MDL Number:
MFCD00173729 CAS Number: 68282-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...