3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Fluorophenyl)pyrazole-4-carboxaldehyde

A401960-1G A401960-25G A401960-5G
77278-740EA 18.54 USD
77278-740 77278-742 77278-744
3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde 98%
3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇FN₂O
MW: 190.18 g/mol
MDL Number: MFCD01922131
CAS Number: 306936-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR