EXP-3174 97%

Supplier: Ambeed
Warning

Synonyms: Losartan carboxylic acid, 1-((2'-(1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-butyl-4-chloro-1H-imidazole-5-carboxylic acid

A401868-1MG A401868-5MG A401868-250MG
77264-528EA 7.21 USD
77264-528 77264-530 77616-738
EXP-3174 97%
EXP-3174

Precaución: for Research Use Only

Formula: C₂₂H₂₁ClN₆O₂
MW: 436.9 g/mol
MDL Number: MFCD00871928
CAS Number: 124750-92-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR