(2-Hydroxy-3-methylphenyl)boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-3-methylphenylboronic acid

A401788-1G A401788-250MG A401788-5G
77278-734EA 49.44 USD
77278-734 77278-736 77278-738
(2-Hydroxy-3-methylphenyl)boronic acid 97%
(2-Hydroxy-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₉BO₃
MW: 151.96 g/mol
MDL Number: MFCD08689549
CAS Number: 259209-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR