(4S,5R)-4-(Fluoromethyl)-5-(4-iodophenyl)-2-phenyl-4,5-dihydrooxazole 97%

Supplier: Ambeed
Warning

A401354-250MG A401354-5G
77240-626EA 13.39 USD
77240-626 77240-628
(4S,5R)-4-(Fluoromethyl)-5-(4-iodophenyl)-2-phenyl-4,5-dihydrooxazole 97%
(4S,5R)-4-(Fluoromethyl)-5-(4-iodophenyl)-2-phenyl-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₁₆H₁₃FINO
MW: 381.18 g/mol
MDL Number: MFCD27977270
CAS Number: 927689-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR