1-Benzyl-N-carbobenzoxy-L-glutamate 97%

Supplier: Ambeed
Warning

Synonyms: N-Cbz-L-glutamic Acid 1-Benzyl Ester, N-Benzyloxycarbonyl-L-glutamic acid 1-benzyl ester, Z-Glu-OBzl, 1-Benzyl N-Cbz-L-glutamate, N-Carbobenzoxy-L-glutamic Acid 1-Benzyl Ester

A400307-100G A400307-10G A400307-1G A400307-25G A400307-5G
77278-718EA 118 USD
77278-718 77278-720 77278-722 77278-724 77278-726
1-Benzyl-N-carbobenzoxy-L-glutamate 97%
1-Benzyl-N-carbobenzoxy-L-glutamate

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₆
MDL Number: MFCD00077013
CAS Number: 3705-42-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR