(4S,4'S)-2,2'-[2-Phenyl-1-(phenylmethyl)ethylidene]bis[4-(1-methylethyl)-4,5-dihydrooxazole 98%

Supplier: Ambeed
Warning

A400037-100MG A400037-1G A400037-250MG
77252-114EA 27 USD
77252-114 77252-116 77252-118
(4S,4'S)-2,2'-[2-Phenyl-1-(phenylmethyl)ethylidene]bis[4-(1-methylethyl)-4,5-dihydrooxazole 98%
(4S,4'S)-2,2'-[2-Phenyl-1-(phenylmethyl)ethylidene]bis[4-(1-methylethyl)-4,5-dihydrooxazole

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₂O₂
MW: 418.57 g/mol
MDL Number: MFCD07368373
CAS Number: 444575-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR