Mycophenolic acid 99+%

Supplier: Ambeed
Warning

Synonyms: (E)-6-(4-Hydroxy-6-methoxy-7-methyl-3-oxo-1,3-dihydroisobenzofuran-5-yl)-4-methylhex-4-enoic acid

A399663-100G A399663-100MG A399663-10G A399663-1G A399663-500G A399663-5G A399663-25G
77291-124EA 87.55 USD
77291-124 77291-126 77291-128 77291-130 77291-132 77291-134 77572-236
Mycophenolic acid 99+%
Mycophenolic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₀O₆
MW: 320.34 g/mol
MDL Number: MFCD00036814
CAS Number: 24280-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR