TAK-700 98%

Supplier: Ambeed
Warning

Synonyms: Orteronel, (S)-6-(7-Hydroxy-6,7-dihydro-5H-pyrrolo[1,2-c]imidazol-7-yl)-N-methyl-2-naphthamide, 6-(7-Hydroxy-6,7-dihydro-5H-pyrrolo[1,2-c]imidazol-7-yl)-N-methylnaphthalene-2-carboxamide

A399493-1MG A399493-5MG
77240-606EA 36.05 USD
77240-606 77240-608
TAK-700 98%
TAK-700

Precaución: for Research Use Only

Formula: C₁₈H₁₇N₃O₂
MW: 307.35 g/mol
MDL Number: MFCD20528078
CAS Number: 566939-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR