2-Bromo-N,N-dimethylbenzamide 98%

Supplier: Ambeed
Warning

A399145-1G A399145-25G A399145-5G
77264-504EA 17.51 USD
77264-504 77264-506 77264-508
2-Bromo-N,N-dimethylbenzamide 98%
2-Bromo-N,N-dimethylbenzamide

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO
MW: 228.0900 g/mol
MDL Number: MFCD00463635
CAS Number: 54616-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR