CGS 21680 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-(4-(2-((6-Amino-9-((2R,3R,4S,5S)-5-(ethylcarbamoyl)-3,4-dihydroxytetrahydrofuran-2-yl)-9H-purin-2-yl)amino)ethyl)phenyl)propanoic acid hydrochloride

A398047-100MG A398047-10MG A398047-1MG A398047-50MG A398047-5MG
77252-094EA 284.28 USD
77252-094 77252-096 77252-098 77252-100 77252-102
CGS 21680 hydrochloride 98%
CGS 21680 hydrochloride

Precaución: for Research Use Only

Formula: C₂₃H₂₉N₇O₆·HCl
MW: 535.98 g/mol
MDL Number: MFCD00078573
CAS Number: 124431-80-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR