Methyl 2-methyl-1H-pyrrole-3-carboxylate 95%

Supplier: Ambeed
Warning

A397484-1G A397484-5G
77264-494EA 84 USD
77264-494 77594-214
Methyl 2-methyl-1H-pyrrole-3-carboxylate 95%
Methyl 2-methyl-1H-pyrrole-3-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉NO₂
MDL Number: MFCD09033483
CAS Number: 3168-85-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR